N-(3,5-Dichlorophenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039371-500MG
101847-358EA 193.85 USD
101847-358
N-(3,5-Dichlorophenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-(3,5-Dichlorophenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₁H₈Cl₂N₂O₂S₂
MDL Number: MFCD12027856
CAS Number: 1142200-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR