N-(3-Chloro-4-methylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Supplier: Matrix Scientific

039365-500MG
101847-346EA 144.9 USD
101847-346
N-(3-Chloro-4-methylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide
N-(3-Chloro-4-methylphenyl)-2-(2-mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)acetamide

Hazard: Irritant

Formula: C₁₂H₁₁ClN₂O₂S₂
MDL Number: MFCD12027850
CAS Number: 1142200-21-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR