2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-phenylacetamide

Supplier: Matrix Scientific

039354-500MG
101847-324EA 193.85 USD
101847-324
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-phenylacetamide
2-(2-Mercapto-4-oxo-4,5-dihydro-1,3-thiazol-5-yl)-N-phenylacetamide

Hazard: Irritant

Formula: C₁₁H₁₀N₂O₂S₂
MDL Number: MFCD12027839
CAS Number: 1142200-00-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR