4,5-Bis(4-methylphenyl)-4H-imidazole-2-thiol

Supplier: Matrix Scientific

039352-500MG
101847-320EA 217.35 USD
101847-320
4,5-Bis(4-methylphenyl)-4H-imidazole-2-thiol
4,5-Bis(4-methylphenyl)-4H-imidazole-2-thiol

Hazard: Irritant

Formula: C₁₇H₁₆N₂S
MDL Number: MFCD12027838
CAS Number: 73181-95-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR