(4E)-3-(Chloromethyl)-4-[(2-methyl-1H-indol-3-yl)methylene]isoxazol-5(4H)-one
Supplier: Matrix Scientific
039342-500MG
101847-300EA
193.85
USD
101847-300
(4E)-3-(Chloromethyl)-4-[(2-methyl-1H-indol-3-yl)methylene]isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-[(2-methyl-1H-indol-3-yl)methylene]isoxazol-5(4H)-one
Hazard: Irritant
Formula:
C₁₄H₁₁ClN₂O₂ |
MDL Number:
MFCD12027829 CAS Number: 1142199-82-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...