(4E)-3-(Chloromethyl)-4-(1H-indol-5-ylmethylene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039340-500MG
101847-296EA 144.9 USD
101847-296
(4E)-3-(Chloromethyl)-4-(1H-indol-5-ylmethylene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(1H-indol-5-ylmethylene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₃H₉ClN₂O₂
MDL Number: MFCD12027827
CAS Number: 1142199-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR