(4E)-3-(Chloromethyl)-4-[(1-methyl-1H-pyrrol-2-yl)methylene]isoxazol-5(4H)-one

Supplier: Matrix Scientific

039339-500MG
101847-294EA 144.9 USD
101847-294
(4E)-3-(Chloromethyl)-4-[(1-methyl-1H-pyrrol-2-yl)methylene]isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-[(1-methyl-1H-pyrrol-2-yl)methylene]isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₀H₉ClN₂O₂
MDL Number: MFCD12027826
CAS Number: 1142199-77-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR