(4E)-3-(Chloromethyl)-4-(3,4-dihydroxybenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039338-500MG
101847-292EA 193.85 USD
101847-292
(4E)-3-(Chloromethyl)-4-(3,4-dihydroxybenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(3,4-dihydroxybenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₈ClNO₄
MDL Number: MFCD12027825
CAS Number: 1142199-74-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR