(4E)-3-(Chloromethyl)-4-(pyridin-2-ylmethylene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039337-500MG
101847-290EA 193.85 USD
101847-290
(4E)-3-(Chloromethyl)-4-(pyridin-2-ylmethylene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(pyridin-2-ylmethylene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₀H₇ClN₂O₂
MDL Number: MFCD12027824
CAS Number: 1142199-73-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR