(4E)-3-(Chloromethyl)-4-(4-hydroxybenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039336-500MG
101847-288EA 193.85 USD
101847-288
(4E)-3-(Chloromethyl)-4-(4-hydroxybenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(4-hydroxybenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₈ClNO₃
MDL Number: MFCD12027823
CAS Number: 1142199-72-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR