(4E)-3-(Chloromethyl)-4-[(3-methyl-2-thienyl)methylene]isoxazol-5(4H)-one

Supplier: Matrix Scientific

039335-500MG
101847-286EA 144.9 USD
101847-286
(4E)-3-(Chloromethyl)-4-[(3-methyl-2-thienyl)methylene]isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-[(3-methyl-2-thienyl)methylene]isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₀H₈ClNO₂S
MDL Number: MFCD12027822
CAS Number: 1142199-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR