(4E)-4-[(5-Bromo-2-furyl)methylene]-3-(chloromethyl)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039334-500MG
101847-284EA 144.9 USD
101847-284
(4E)-4-[(5-Bromo-2-furyl)methylene]-3-(chloromethyl)isoxazol-5(4H)-one
(4E)-4-[(5-Bromo-2-furyl)methylene]-3-(chloromethyl)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₉H₅BrClNO₃
MDL Number: MFCD12027821
CAS Number: 1142199-66-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR