(4E)-3-(Chloromethyl)-4-[(5-methyl-2-furyl)methylene]isoxazol-5(4H)-one

Supplier: Matrix Scientific

039333-500MG
101847-282EA 193.85 USD
101847-282
(4E)-3-(Chloromethyl)-4-[(5-methyl-2-furyl)methylene]isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-[(5-methyl-2-furyl)methylene]isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₀H₈ClNO₃
MDL Number: MFCD12027820
CAS Number: 1142199-65-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR