(4E)-4-(2,1,3-Benzothiadiazol-4-ylmethylene)-3-(chloromethyl)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039332-500MG
101847-280EA 193.85 USD
101847-280
(4E)-4-(2,1,3-Benzothiadiazol-4-ylmethylene)-3-(chloromethyl)isoxazol-5(4H)-one
(4E)-4-(2,1,3-Benzothiadiazol-4-ylmethylene)-3-(chloromethyl)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₆ClN₃O₂S
MDL Number: MFCD12027819
CAS Number: 1142199-64-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR