(4E)-4-(4-Bromobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039326-500MG
101847-268EA 193.85 USD
101847-268
(4E)-4-(4-Bromobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one
(4E)-4-(4-Bromobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₇BrClNO₂
MDL Number: MFCD12027813
CAS Number: 1142199-54-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR