(4E)-4-(3-Bromobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039325-500MG
101847-266EA 144.9 USD
101847-266
(4E)-4-(3-Bromobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one
(4E)-4-(3-Bromobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₇BrClNO₂
MDL Number: MFCD12027812
CAS Number: 1142199-51-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR