(4E)-3-(Chloromethyl)-4-(2,4,5-trimethoxybenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039322-500MG
101847-260EA 193.85 USD
101847-260
(4E)-3-(Chloromethyl)-4-(2,4,5-trimethoxybenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(2,4,5-trimethoxybenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₄H₁₄ClNO₅
MDL Number: MFCD12027809
CAS Number: 1142199-46-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR