(4E)-3-(Chloromethyl)-4-(4-methylbenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039321-500MG
101847-258EA 144.9 USD
101847-258
(4E)-3-(Chloromethyl)-4-(4-methylbenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(4-methylbenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₂
MDL Number: MFCD12027808
CAS Number: 1142199-45-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR