(4E)-3-(Chloromethyl)-4-(2-fluorobenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039309-500MG
101847-234EA 144.9 USD
101847-234
(4E)-3-(Chloromethyl)-4-(2-fluorobenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(2-fluorobenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₇ClFNO₂
MDL Number: MFCD12027796
CAS Number: 1142199-21-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR