(4E)-3-(Chloromethyl)-4-(4-ethoxybenzylidene)isoxazol-5(4H)-one
Supplier: Matrix Scientific
039308-500MG
101847-232EA
193.85
USD
101847-232
(4E)-3-(Chloromethyl)-4-(4-ethoxybenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(4-ethoxybenzylidene)isoxazol-5(4H)-one
Hazard: Irritant
Formula:
C₁₃H₁₂ClNO₃ |
MDL Number:
MFCD12027795 CAS Number: 1142199-20-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...