(4E)-3-(Chloromethyl)-4-(3-methoxybenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039306-500MG
101847-228EA 144.9 USD
101847-228
(4E)-3-(Chloromethyl)-4-(3-methoxybenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(3-methoxybenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₂H₁₀ClNO₃
MDL Number: MFCD12027793
CAS Number: 1142199-16-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR