(4E)-3-(Chloromethyl)-4-[4-(dimethylamino)benzylidene]isoxazol-5(4H)-one

Supplier: Matrix Scientific

039303-500MG
101847-222EA 144.9 USD
101847-222
(4E)-3-(Chloromethyl)-4-[4-(dimethylamino)benzylidene]isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-[4-(dimethylamino)benzylidene]isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₃H₁₃ClN₂O₂
MDL Number: MFCD12027790
CAS Number: 1142199-10-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR