(4E)-3-(Chloromethyl)-4-(2,5-dimethoxybenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039301-500MG
101847-218EA 193.85 USD
101847-218
(4E)-3-(Chloromethyl)-4-(2,5-dimethoxybenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(2,5-dimethoxybenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₃H₁₂ClNO₄
MDL Number: MFCD12027788
CAS Number: 1142199-06-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR