(4E)-3-(Chloromethyl)-4-(2,4-dichlorobenzylidene)isoxazol-5(4H)-one
Supplier: Matrix Scientific
039297-500MG
101847-210EA
193.85
USD
101847-210
(4E)-3-(Chloromethyl)-4-(2,4-dichlorobenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(2,4-dichlorobenzylidene)isoxazol-5(4H)-one
Hazard: Irritant
Formula:
C₁₁H₆Cl₃NO₂ |
MDL Number:
MFCD12027784 CAS Number: 1142199-00-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...