(4E)-3-(Chloromethyl)-4-(2,3-dichlorobenzylidene)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039296-500MG
101847-208EA 193.85 USD
101847-208
(4E)-3-(Chloromethyl)-4-(2,3-dichlorobenzylidene)isoxazol-5(4H)-one
(4E)-3-(Chloromethyl)-4-(2,3-dichlorobenzylidene)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₆Cl₃NO₂
MDL Number: MFCD12027783
CAS Number: 1142198-98-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR