(4E)-4-(4-Chlorobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039294-500MG
101847-204EA 193.85 USD
101847-204
(4E)-4-(4-Chlorobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one
(4E)-4-(4-Chlorobenzylidene)-3-(chloromethyl)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₇Cl₂NO₂
MDL Number: MFCD12027781
CAS Number: 1142198-93-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR