(4E)-4-Benzylidene-3-(chloromethyl)isoxazol-5(4H)-one

Supplier: Matrix Scientific

039290-500MG
101847-196EA 144.9 USD
101847-196
(4E)-4-Benzylidene-3-(chloromethyl)isoxazol-5(4H)-one
(4E)-4-Benzylidene-3-(chloromethyl)isoxazol-5(4H)-one

Hazard: Irritant

Formula: C₁₁H₈ClNO₂
MDL Number: MFCD12027777
CAS Number: 1142198-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR