1-(2,3-Dihydro-1,4-benzodioxin-6-yl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039281-500MG
101847-178EA 144.9 USD
101847-178
1-(2,3-Dihydro-1,4-benzodioxin-6-yl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(2,3-Dihydro-1,4-benzodioxin-6-yl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₄H₁₆N₂O₂S
MDL Number: MFCD12027768
CAS Number: 1142213-30-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR