1-[4-(1,3-Benzoxazol-2-yl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039279-500MG
101847-174EA 193.85 USD
101847-174
1-[4-(1,3-Benzoxazol-2-yl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-[4-(1,3-Benzoxazol-2-yl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₉H₁₇N₃OS
MDL Number: MFCD12027766
CAS Number: 1142213-27-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR