2-Hydroxy-5-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid

Supplier: Matrix Scientific

039278-500MG
101847-172EA 144.9 USD
101847-172
2-Hydroxy-5-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid
2-Hydroxy-5-(2-mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzoic acid

Hazard: Irritant

Formula: C₁₃H₁₄N₂O₃S
MDL Number: MFCD12027765
CAS Number: 1142213-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR