4-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzamide

Supplier: Matrix Scientific

039275-500MG
101847-166EA 144.9 USD
101847-166
4-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzamide
4-(2-Mercapto-4,4-dimethylpyrimidin-1(4H)-yl)benzamide

Hazard: Irritant

Formula: C₁₃H₁₅N₃OS
MDL Number: MFCD12027762
CAS Number: 1142213-19-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR