4,4-Dimethyl-1-[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039274-500MG
101847-164EA 193.85 USD
101847-164
4,4-Dimethyl-1-[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]-1,4-dihydropyrimidine-2-thiol
4,4-Dimethyl-1-[2,3,5,6-tetrafluoro-4-(trifluoromethyl)phenyl]-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₃H₉F₇N₂S
MDL Number: MFCD12027761
CAS Number: 1142213-18-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR