1-(2,4-Dimethoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039271-500MG
101847-158EA 144.9 USD
101847-158
1-(2,4-Dimethoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(2,4-Dimethoxyphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₂S
MDL Number: MFCD12027758
CAS Number: 1142212-70-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR