1-(4-Chloro-2-methoxy-5-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039264-500MG
101847-144EA 193.85 USD
101847-144
1-(4-Chloro-2-methoxy-5-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(4-Chloro-2-methoxy-5-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₄H₁₇ClN₂OS
MDL Number: MFCD12027751
CAS Number: 1142212-57-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR