1-[2-Chloro-5-(trifluoromethyl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039263-500MG
101847-142EA 193.85 USD
101847-142
1-[2-Chloro-5-(trifluoromethyl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-[2-Chloro-5-(trifluoromethyl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₃H₁₂ClF₃N₂S
MDL Number: MFCD12027750
CAS Number: 1142212-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR