1-[4-Chloro-2-(trifluoromethyl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
Supplier: Matrix Scientific
039262-500MG
101847-140EA
193.85
USD
101847-140
1-[4-Chloro-2-(trifluoromethyl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-[4-Chloro-2-(trifluoromethyl)phenyl]-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
Hazard: Irritant
Formula:
C₁₃H₁₂ClF₃N₂S |
MDL Number:
MFCD12027749 CAS Number: 1142212-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...