4,4-Dimethyl-1-(4-methylphenyl)-1,4-dihydropyrimidine-2-thiol
Supplier: Matrix Scientific
039261-500MG
101847-138EA
144.9
USD
101847-138
4,4-Dimethyl-1-(4-methylphenyl)-1,4-dihydropyrimidine-2-thiol
4,4-Dimethyl-1-(4-methylphenyl)-1,4-dihydropyrimidine-2-thiol
Hazard: Irritant
Formula:
C₁₃H₁₆N₂S |
MDL Number:
MFCD12027748 CAS Number: 1142212-52-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...