1-(2,4-Dimethylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039254-500MG
101847-124EA 193.85 USD
101847-124
1-(2,4-Dimethylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(2,4-Dimethylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₄H₁₈N₂S
MDL Number: MFCD12027741
CAS Number: 1142212-39-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR