1-(3,5-Dichlorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039252-500MG
101847-120EA 193.85 USD
101847-120
1-(3,5-Dichlorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(3,5-Dichlorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₂H₁₂Cl₂N₂S
MDL Number: MFCD12027739
CAS Number: 1142212-36-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR