1-(2,5-Dichlorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039250-500MG
101847-116EA 193.85 USD
101847-116
1-(2,5-Dichlorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(2,5-Dichlorophenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₂H₁₂Cl₂N₂S
MDL Number: MFCD12027737
CAS Number: 1142212-32-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR