1-(5-Chloro-2-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
Supplier: Matrix Scientific
039247-500MG
101847-110EA
193.85
USD
101847-110
1-(5-Chloro-2-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(5-Chloro-2-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
Hazard: Irritant
Formula:
C₁₃H₁₅ClN₂S |
MDL Number:
MFCD12027734 CAS Number: 1142212-27-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...