1-(3-Chloro-4-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Supplier: Matrix Scientific

039246-500MG
101847-108EA 193.85 USD
101847-108
1-(3-Chloro-4-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol
1-(3-Chloro-4-methylphenyl)-4,4-dimethyl-1,4-dihydropyrimidine-2-thiol

Hazard: Irritant

Formula: C₁₃H₁₅ClN₂S
MDL Number: MFCD12027733
CAS Number: 1142212-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR