4,4-Dimethyl-1-phenyl-1,4-dihydropyrimidine-2-thiol
Supplier: Matrix Scientific
039235-500MG
101847-086EA
144.9
USD
101847-086
4,4-Dimethyl-1-phenyl-1,4-dihydropyrimidine-2-thiol
4,4-Dimethyl-1-phenyl-1,4-dihydropyrimidine-2-thiol
Hazard: Irritant
Formula:
C₁₂H₁₄N₂S |
MDL Number:
MFCD12027722 CAS Number: 146496-45-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...