4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzamide

Supplier: Matrix Scientific

039233-500MG
101847-082EA 290.77 USD
101847-082
4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzamide
4-Amino-N-(5-methyl-1,3,4-thiadiazol-2-yl)benzamide

Hazard: Irritant

Formula: C₁₀H₁₀N₄OS
MW: 234.28 g/mol
MDL Number: MFCD01666707
CAS Number: 36855-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR