6-Chloro-8-nitro-4-oxo-4H-chromene-3-carbaldehyde

Supplier: Matrix Scientific

039230-500MG
101847-076EA 217.35 USD
101847-076
6-Chloro-8-nitro-4-oxo-4H-chromene-3-carbaldehyde
6-Chloro-8-nitro-4-oxo-4H-chromene-3-carbaldehyde

Hazard: Irritant

Formula: C₁₀H₄ClNO₅
MDL Number: MFCD11899892
CAS Number: 879559-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR