3-Formyl-6-methylchromone

Supplier: Matrix Scientific

Synonyms: 6-Methyl-4-oxo-4H-chromene-3-carbaldehyde

039229-500MG
101847-074EA 290.77 USD
101847-074
3-Formyl-6-methylchromone
3-Formyl-6-methylchromone

Hazard: Irritant

Formula: C₁₁H₈O₃
MW: 188.1794 g/mol
MDL Number: MFCD00138943
CAS Number: 42059-81-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR