2-Amino-3-formyl-6-methylchromone

Supplier: Matrix Scientific

039222-500MG
101847-060EA 217.35 USD
101847-060
2-Amino-3-formyl-6-methylchromone
2-Amino-3-formyl-6-methylchromone

Hazard: Irritant

Formula: C₁₁H₉NO₃
MDL Number: MFCD00138945
CAS Number: 68301-75-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR