2-Amino-6,8-dimethyl-4-oxo-4H-chromene-3-carbaldehyde

Supplier: Matrix Scientific

039219-500MG
101847-054EA 290.77 USD
101847-054
2-Amino-6,8-dimethyl-4-oxo-4H-chromene-3-carbaldehyde
2-Amino-6,8-dimethyl-4-oxo-4H-chromene-3-carbaldehyde

Hazard: Irritant

Formula: C₁₂H₁₁NO₃
MDL Number: MFCD09261020
CAS Number: 68301-79-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR