[[2-(4-Hydroxypiperidin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid

Supplier: Matrix Scientific

039207-500MG
101847-032EA 144.9 USD
101847-032
[[2-(4-Hydroxypiperidin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid
[[2-(4-Hydroxypiperidin-1-yl)-2-oxoethyl](phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₅H₂₀N₂O₄
MDL Number: MFCD12027710
CAS Number: 1142211-90-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR