[(2-Oxo-2-thiomorpholin-4-ylethyl)(phenyl)amino]acetic acid

Supplier: Matrix Scientific

039201-500MG
101847-020EA 144.9 USD
101847-020
[(2-Oxo-2-thiomorpholin-4-ylethyl)(phenyl)amino]acetic acid
[(2-Oxo-2-thiomorpholin-4-ylethyl)(phenyl)amino]acetic acid

Hazard: Irritant

Formula: C₁₄H₁₈N₂O₃S
MDL Number: MFCD12027704
CAS Number: 1142211-78-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR